Learn More
Eisco™ Oligoclase Mineral Specimen
SCI_ED
Catalog No. S27604 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27604 12/Pk.
S27582 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27604 Supplier Eisco™ Supplier No. ESNG0075PK12
Add to Cart
Add to Cart
Oligoclase is a feldspar with high levels of sodium.
  • Correlates well to Next Generation Science Standards Evidence
  • Standard hardness of 6
  • Exhibits two planes of cleavage
  • Perfect size for use in classroom identification labs and exercises
Specifications
Product Type Mineral
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm