Learn More
Eisco™ Oligoclase Mineral Specimen
SCI_ED
Catalog No. S26662 Shop All Eisco Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S26662 10/Pk.
1 options
Catalog No. S26662 Supplier Eisco™ Supplier No. ESNG0075PK10
Add to Cart
Add to Cart
Oligoclase is a plagioclase fledspar that exemplifies many mineral physical properties including hardness, cleavage, and luster.
  • A plagioclase feldspar with high levels of sodium
  • Standard hardness of 6 on Mohs scale
  • Exhibits two planes of cleavage
  • Characteristic nonmetallic luster
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Ideal for learning about mineralogy or igneous petrology
  • Great addition to any identification exercises, especially when paired with an alkali feldspar
Specifications
Product Type Metamorphic Rock
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Oligoclase