Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oligodendrocyte Marker O1 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1327U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1327U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1327U100 Supplier R&D Systems Supplier No. FAB1327U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Oligodendrocyte Marker O1 Monoclonal specifically detects Oligodendrocyte Marker O1 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry.

Specifications

Antigen Oligodendrocyte Marker O1
Applications Flow Cytometry
Classification Monoclonal
Clone O1
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Host Species Mouse
Immunogen Bovine brain corpus callosum white matter
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human, mouse, rat and chicken Oligodendrocyte Marker O1.
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgM
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.