Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oligodendrocyte Marker O4 NorthernLights™ NL493-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems NL1326G
This item is not returnable.
View return policy
Description
Oligodendrocyte Marker O4 Monoclonal specifically detects Oligodendrocyte Marker O4 in Human, Mouse, Rat, Chicken samples. It is validated for Immunocytochemistry.Specifications
| Oligodendrocyte Marker O4 | |
| Monoclonal | |
| NorthernLights 493 (NL493) | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Bovine brain corpus callosum white matter. | |
| 0.5 mL | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| IgM |
| Immunocytochemistry | |
| O4 | |
| Immunocytochemistry | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Detects human, mouse, rat, and chicken Oligodendrocyte Marker O4. | |
| Store the unopened product at 2 - 8degrees C. Do not use past expiration date. Protect from light. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction