Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oligomycin, >98%, For TLC analysis, MP Biomedicals™

Catalog No. ICN15178610 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15178610 10 mg
ICN15178601 1 mg
ICN15178605 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15178610 Supplier MP Biomedicals, Inc Supplier No. 0215178610

Chemical Identifiers

CAS 1404-19-9
Molecular Formula C45H74O11
Molecular Weight (g/mol) 791.08
MDL Number MFCD00065705,MFCD01779388
InChI Key MNULEGDCPYONBU-ZUSSGZTJNA-N
Synonym Olivomycin, Aburamycin
PubChem CID 78358496
IUPAC Name (4E,18E,20Z)-22-ethyl-7,11,14,15-tetrahydroxy-6'-(2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-2,26-dioxaspiro[bicyclo[23.3.1]nonacosane-27,2'-oxane]-4,18,20-triene-3,9,13-trione
SMILES CCC1CCC2OC3(CCC(C)C(CC(C)O)O3)C(C)C(OC(=O)\C=C\C(C)C(O)C(C)C(=O)C(C)C(O)C(C)C(=O)C(C)(O)C(O)C(C)C\C=C\C=C/1)C2C

Specifications

Type Oligomycin
Physical Form Powder
Color White
Quantity 10 mg
Formula Weight ≈789.3
Percent Purity >98%
Chemical Name or Material Oligomycin, For TLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.