Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olive oil, pure, virgin

Catalog No. p-4323719
Click to view available options
Quantity:
25 mL
500 mL
2.5 L
Packaging:
Glass bottle

CAS: 8001-25-0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-25-0

Specifications

Acidity 2.0mg KOH/g max.
Absorbance 0.2 (270nm, 1g/100ml cyclohexane) max. at 425nm
Color Yellow
Density 0.9135g/mL
Flash Point 225°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4670 to 1.4710
Quantity 25 mL
Saponification Value 184 to 196mg KOH/g
Merck Index 14,6836
Specific Gravity 0.9135
Solubility Information Solubility in water: .. Other solubilities: practically insoluble in alcohol, miscible with light petroleum (50-60°C)
Grade Pure
Concentration or Composition (by Analyte or Components) of fatty acids: 0.1 % max. (< C16) 7.5 to 20.0 % (Palmitic acid) 3.5 % max. (Palmitoleic acid) 0.5 to 5.0 % (Stearic acid) 56.0 to 85.0 % (Oleic acid) 3.5 to 20.0 % (Linoleic acid) 1.2 % max. (Linolenic acid) 0.7 % max. (Arachidic acid) 0.4 % max. (Eic
Physical Form Powder
Chemical Name or Material Olive oil
Show More Show Less
EINECSNumber 232-277-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.