Learn More
Eisco™ Olivine Mineral Specimen
SCI_ED
Catalog No. S24706 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24706 1 Ea. 2 to 3 cm
S26631 10/Pk. 2 to 3 cm
S24707 12/Pk. 2 to 3 cm
S26648 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24706 Supplier Eisco™ Supplier No. ESNG0023
Add to Cart
Add to Cart
Olivine Mineral Specimen for teaching students about mineral physical properties like hardness, fracture, and luster.
  • For use in lessons on Bowen's reaction series
  • First mineral to crystallize in cooling magma
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Massive earthy habit is useful example of nonmetallic luster
Specifications
Product Type Mineral
For Use With (Application) Study of mineral physical properties like hardness, fracture, and luster
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Specimen Type Olivine