Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olomoucine, 98%, high purity, Thermo Scientific™

Catalog No. AC328410010
Click to view available options
:
1mg

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 101622-51-9
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C15H18N6O
Quantity 1mg
MDL Number MFCD00189360
Synonym olomoucine, 4erk, unii-6a839b2hys, insolution olomoucine, 2-2-hydroxyethylamino-6-benzylamino-9-methylpurine, 6-benzylamino-2-2-hydroxyethylamino-9-methylpurine, 2-6-benzylamino-9-methyl-9h-purin-2-yl amino ethanol, ethanol,2-9-methyl-6-phenylmethyl amino-9h-purin-2-yl amino, 2-hydroxyethylamino-6-benzylamino-9-methylpurine
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: soluble in dmso and methanol
InChI Key GTVPOLSIJWJJNY-UHFFFAOYSA-N
SMILES CN1C=NC2=C(NCC3=CC=CC=C3)N=C(NCCO)N=C12
IUPAC Name 2-{[6-(benzylamino)-9-methyl-9H-purin-2-yl]amino}ethan-1-ol
Molecular Weight (g/mol) 298.35
PubChem CID 4592
Formula Weight 298.4
Percent Purity 98%
Chemical Name or Material Olomoucine, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.