Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OLT-2 Antibody (2564C) [Janelia Fluor™ 646], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10059J
Description
OLT-2 Monoclonal antibody specifically detects OLT-2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
OLT-2 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from cell culture supernatant | |
RUO | |
441864 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, CyTOF | |
2564C | |
Flow Cytometry, CyTOF-ready | |
OSCAR-Like Transcript-2 Protein, T Cell-Interacting, Activating Receptor On Myeloid Cells, T Cell-Interacting, Activating Receptor On Myeloid Cells 1, TARM1, T-Cell-Interacting, Activating Receptor On Myeloid Cells Protein 1 | |
Chinese hamster ovary cell line CHO-derived recombinant human OLT-2/TARM-1, Met1-Val235, Accession # B6A8C7 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction