Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OMA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598794
Description
OMA1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
OMA1 (Overlapping with the m-AAA protease 1 homolog) is also named as MPRP1(Metalloprotease-related protein 1) and belongs to the peptidase M48 family. It is a zinc metalloprotease that spans the inner mitochondrial membrane with a number of predicted membrane spanning domains and the 60 kDa form of OMA1 rapidly accumulated concomitantly with the cleavage of all OPA1 variants, and the 40 kDa form of OMA1 may be the inactive form, with the 60 kDa form being the active enzyme.
Specifications
OMA1 | |
Polyclonal | |
Unconjugated | |
OMA1 | |
2010001O09Rik; DAB1; metalloendopeptidase OMA1, mitochondrial; metalloprotease related protein 1; metalloprotease-related protein 1; MPRP1; MPRP-1; Oma1; OMA1 homolog, zinc metallopeptidase; OMA1 homolog, zinc metallopeptidase (S. cerevisiae); OMA1 zinc metallopeptidase; OMA1 zinc metallopeptidase homolog; overlapping activity with M-AAA protease; overlapping with the m-AAA protease 1 homolog; peptidase; RGD1304821; YKR087C; zinc metallopeptidase homolog; zinc metallopeptidase OMA1; ZMPOMA1 | |
Rabbit | |
Protein G | |
RUO | |
115209 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q96E52 | |
OMA1 | |
Recombinant Human Metalloendopeptidase OMA1, mitochondrial protein (22-194AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction