Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OMA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106421 Supplier Invitrogen™ Supplier No. PA5106421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OMA1.

OMA1 (Overlapping with the m-AAA protease 1 homolog) is also named as MPRP1(Metalloprotease-related protein 1) and belongs to the peptidase M48 family. It is a zinc metalloprotease that spans the inner mitochondrial membrane with a number of predicted membrane spanning domains and the 60 kDa form of OMA1 rapidly accumulated concomitantly with the cleavage of all OPA1 variants, and the 40 kDa form of OMA1 may be the inactive form, with the 60 kDa form being the active enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OMA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OMA1
Gene Accession No. D3ZS74, Q96E52, Q9D8H7
Gene Alias 2010001O09Rik; DAB1; metalloendopeptidase OMA1, mitochondrial; metalloprotease related protein 1; metalloprotease-related protein 1; MPRP1; MPRP-1; Oma1; OMA1 homolog, zinc metallopeptidase; OMA1 homolog, zinc metallopeptidase (S. cerevisiae); OMA1 zinc metallopeptidase; OMA1 zinc metallopeptidase homolog; overlapping activity with M-AAA protease; overlapping with the m-AAA protease 1 homolog; peptidase; RGD1304821; YKR087C; zinc metallopeptidase homolog; zinc metallopeptidase OMA1; ZMPOMA1
Gene Symbols OMA1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OMA1(Accession Q96E52), corresponding to amino acid residues K405-Q455.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115209, 298282, 67013
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.