Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Omega-hexatoxin-Hv1a Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144389 Supplier Invitrogen™ Supplier No. PA5144389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Omega-hexatoxin-Hv1a Polyclonal Antibody for Western Blot, ELISA

Reversibly and voltage-independently blocks both mid-low- (M-LVA) and high-voltage-activated (HVA) calcium channels in cockroach DUM neurons (PubMed:17610847). Is lethal to many insect species but not toxic to mammals (PubMed:9228949, PubMed:16779650). May target the insect high-voltage-activated calcium channel Dmca1D. Also inhibits acarines calcium channels. An extremely high toxin concentration partially inhibits Cav1.2/CACNA1C, Cav2.1/CACNA1A and Cav2.2/CACNA1B calcium channel of rats. As for omega-AcTx-Hv2a, the phenotypic effect of injection of this toxin into lone star ticks (Amblyomma americanum) is curling of all eight legs into closed loops.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Omega-hexatoxin-Hv1a
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Omega-hexatoxin-Hv1a
Host Species Rabbit
Immunogen Recombinant Hadronyche versuta Omega-hexatoxin-Hv1a protein (1-37aa ).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Arthropod
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.