Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OMG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552972
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552972 Supplier Invitrogen™ Supplier No. PA552972
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPSGLQENII HLNLSYNHFT DLHNQLTQYT NLRTLDISNN RLESLPAHLP RSLWNMSAAN NNIKLLDKSD TAYQWNLKYL DVSKNMLEKV VLIKNTLRSL EVLNLSSNK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OMG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OMG
Gene Accession No. P23515
Gene Alias oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; OMGP
Gene Symbols OMG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OMG. Recombinant protein control fragment (Product #RP-89910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4974
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.