Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OMG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521750
Description
Recommended positive controls: A549, IMR32, HCT116. Predicted reactivity: Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.
Specifications
OMG | |
Polyclonal | |
Unconjugated | |
OMG | |
oligodendrocyte myelin glycoprotein; oligodendrocyte-myelin glycoprotein; Omg; OMGP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18377, 4974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
1 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
P23515, Q63912 | |
OMG | |
Synthetic peptide corresponding to a region within amino acids 249 and 341 of OMG (Uniprot ID#P23515). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction