Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OMG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 127011AP150UL

Description
Oligodendrocyte myelin glycoprotein (OMG or OMGP) is a cell adhesion molecule contributing to the interactive process required for myelination in the central nervous system. It is also involved in axonogenesis, neuron projection regeneration, and regulation of collateral sprouting of intact axons in response to injury.Specifications
OMG | |
Polyclonal | |
Unconjugated | |
OMG | |
OMG, OMGP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18377, 450224, 4974 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P23515, Q63912 | |
Omg | |
OMG Fusion Protein Ag3210 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction