Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diba Omnifit™ Bottle Waste Cap Thread

Catalog No. 14500712 Shop All Oakton Products
Change view
Click to view available options
Format:
Four Luer Port
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
14-500-712 Four Luer Port
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-500-712 Supplier Diba Supplier No. 2194297
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Connect a vapor trap using the vent port to prevent solvent vapors from escaping

  • Use without any fittings or use male Luer fittings to connect various tubing sizes
  • Anti-twist design

Specifications

Product Type Threaded Closure
Pre-sterilized No
Closure Size 38-430 (GL38)
Liner Material PTFE
Autoclavable Autoclavable
Material Polypropylene
Color Blue
For Use With (Application) Waste disposal
Format Four Luer Port
Height (Metric) 2.54 cm
Quantity 1 Each
Size 38-430 (GL38)
Length (Metric) 5.08 cm
Sterility Non-sterile
For Use With (Equipment) Diba Solvent Waste Bottles
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.