Learn More
Kinesis™ Omnifit™ Gripper Fittings
Catalog No. OF002312 Shop All Oakton Products
Change view
Click to view available options
For Use With (Equipment):
1/16 in. Tubing
1/8 in. Tubing
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
OF002312 1/8 in. Tubing
OF002310 1/16 in. Tubing
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OF002312 Supplier Kinesis™ Supplier No. 002312
Flangeless systems with 1/4 in.-28 United National Fine (UNF) thread provides connection for tubing with an outside diameter of 1/16 in. or 1/8 in.
  • Incorporate a PTFE-seal press-fitted into 316 stainless-steel housing for leak-resistant connection with minimal disturbance to the fluid path
  • Allow repeated connect/disconnect without sealing-face deterioration
  • Gripper fittings do not compress the tubing if over-tightened, and do not twist the tubing during connection
  • Pressure rated to 1000 psi (68 bar)
Use with Omnifit Tube End Fittings
To Use
  • Trim tubing to a point and slide on Tube End fitting
  • Add the Gripper fitting to tubing, ensuring the PTFE seal faces the pointed tubing end
  • Using pliers, pull tubing through Gripper fitting until PTFE seal has reached the uncut portion
  • Rotate Gripper fitting around tubing to seat; trim excess tubing off
Specifications
For Use With (Equipment) 1/8 in. Tubing
Type Gripper Fitting
Max. Pressure 1000 psi