Learn More
Diba Omnifit™ Luer Adapter
Catalog No. OF002501
Change view
Click to view available options
Type:
Female Luer Fitting
Male Luer Fitting
Material:
ETFE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Material
OF002501 Male Luer Fitting ETFE
OF002502 Female Luer Fitting ETFE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OF002501 Supplier Diba Supplier No. 2194010
Adapters fit into 1/4 in.–28 UNF flat bottom ports in most Omnifit connectors and valves
  • Provide quick, push-on connection for softwall tubing or luer
  • Made from chemically resistant ETFE
  • Autoclavable
  • OF001206 features a thread to male Luer lock adapter with a polypropylene lock ring
  • OF002501 features male luer fittings to 1/4 in.–28 UNF thread
  • OF002502 features femal luer fittings to 1/4 in.–28 UNF thread
Specifications
Type Male Luer Fitting
Material ETFE