Description
- Provide quick, push-on connection for softwall tubing or luer
- Made from chemically resistant ETFE
- Autoclavable
- OF001206 features a thread to male Luer lock adapter with a polypropylene lock ring
- OF002501 features male luer fittings to 1/4 in.–28 UNF thread
- OF002502 features femal luer fittings to 1/4 in.–28 UNF thread
Specifications
Specifications
| Type | Female Luer Fitting |
| Material | ETFE |