Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ omp38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144424 Supplier Invitrogen™ Supplier No. PA5144424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

omp38 Polyclonal Antibody for Western Blot, ELISA

Porin. Induces apoptosis in human cells through caspases-dependent and AIF-dependent pathways. Purified Omp38 enters the cells and localizes to the mitochondria, which leads to a release of proapoptotic molecules such as cytochrome c and AIF (apoptosis-inducing factor).
TRUSTED_SUSTAINABILITY

Specifications

Antigen omp38
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias omp38; Outer membrane protein Omp38
Host Species Rabbit
Immunogen Recombinant Acinetobacter baumannii Outer membrane protein Omp38 (omp38) (20-356aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.