Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Oncomodulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598025
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598025 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598025 Supplier Invitrogen™ Supplier No. PA598025
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Oncomodulin Polyclonal Antibody for Western Blot, ELISA

Oncomodulin, encoded by the OCM gene located on chromosome 7p22.3, is a protein that belongs to the parvalbumin family of calcium-binding proteins. It is most prominently found in the inner ear, as well as in certain retinal cells and regenerating neurons. Oncomodulin plays a crucial role in calcium ion homeostasis, which is vital for various cellular processes, including signal transduction, cellular motility, and growth. Notably, oncomodulin has been studied for its role in promoting nerve regeneration, particularly within the central nervous system, where it enhances axonal growth and neuronal repair. This makes it a molecule of significant interest for therapeutic strategies aiming to treat nerve injuries or neurodegenerative diseases. Beyond its neuroregenerative potential, oncomodulin is also expressed in some tumor types, which initially led to its association with cancer-related studies. Current research continues to explore its dual roles in both oncology and neurobiology to better understand its functional implications across different tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oncomodulin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Ocm
Gene Accession No. P51879
Gene Alias hCG18255; Ocm; OCM1; Ocm2; OCMN; OM; ONCM; oncmodulin; oncomodulin; oncomodulin 1; oncomodulin 2; oncomodulin-1; Parvalbumin beta
Gene Symbols Ocm
Host Species Rabbit
Immunogen Recombinant Mouse Oncomodulin protein (2-109AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18261
Target Species Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.