Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oncostatin M/OSM Antibody (17022), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2951I
Description
Oncostatin M/OSM Monoclonal antibody specifically detects Oncostatin M/OSM in Human samples. It is validated for Neutralization, Flow Cytometry, CyTOFSpecifications
Oncostatin M/OSM | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Neutralization, Flow Cytometry, CyTOF | |
17022 | |
Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
MGC20461, oncostatin M, oncostatin-M | |
E. coli-derived recombinant human Oncostatin M/OSM, Ala26-Arg221, Accession # P13725 | |
0.5 mL | |
Biologically Active Proteins, Cell Cycle and Replication | |
5008 | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction