Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oncostatin M/OSM Antibody (OTI2B6), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273146R
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Oncostatin M/OSM | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
OSM | |
Recombinant protein expressed in E.coli corresponding to amino acids 25-252 of human Oncostatin M/OSM. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2B6 | |
Western Blot, Immunohistochemistry | |
MGC20461, oncostatin M, oncostatin-M | |
Mouse | |
Affinity Purified | |
Biologically Active Proteins, Cell Cycle and Replication | |
5008.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction