Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Oncostatin M/OSM Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2951SP
This item is not returnable.
View return policy
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
Oncostatin M/OSM | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC20461, oncostatin M, oncostatin-M | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
5008 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Neutralization, Flow Cytometry, CyTOF | |
17022 | |
Neutralization 0.3-1.5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P13725 | |
OSM | |
E. coli-derived recombinant human Oncostatin M/OSM Ala26-Arg221 Accession # P13725 | |
25 μg | |
Primary | |
Detects human Oncostatin M/OSM in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Oncostatin M, recombinant human (rh) CLC, rhCNTF, rhCardiotrophin-1, rhIL-6, rhIL-11, or rhLIF is observed. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction