Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oncostatin M/OSM Rabbit anti-Human, Clone: 15, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | Oncostatin M/OSM |
|---|---|
| Clone | 15 |
| Dilution | ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 |
| Applications | ELISA, Immunofluorescence |
| Classification | Monoclonal |
Description
Oncostatin M/OSM Monoclonal specifically detects Oncostatin M/OSM in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Oncostatin M/OSM | |
| ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC20461, oncostatin M, oncostatin-M | |
| Recombinant Human Oncostatin M/OSM protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 15 | |
| ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 0.2 um filtered solution in PBS | |
| 5008 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title