Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ondansetron hydrochloride
GREENER_CHOICESDP

Catalog No. 289150 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-915-0 50 mg
28-911-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-915-0 Supplier R&D Systems™ Supplier No. 2891/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective 5-HT3 antagonist

Ondansetron hydrochloride is a selective 5-HT3 receptor antagonist (Ki = 6.16 nM). Also an inhibitor of human multidrug and toxin extrusion (MATE) transporter 1 (IC50 = <10-160 nM). Ondansetron blocks 5-HT-evoked transient inward currents (IC50 = 0.1 nM) in human recombinant h5-HT3A receptors. In an animal model of Parkinson's disease, Ondansetron reduces both the severity of dyskinesia and psychosis-like behaviors. Antiemetic; prevents emesis induced by cytotoxic drugs and radiation.

Chemical Identifiers

CAS 99614-01-4
Molecular Formula C18H20ClN3O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 329.83
Purity >99%
Molecular Weight (g/mol) 329.83
Chemical Name or Material Ondansetron hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.