Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ONE-Glo™ EX Luciferase Assay System

Catalog No. PRE8110 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
10 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE8110 10 mL
PRE8150 10 x 100 mL
PRE8130 10 x 10 mL
PRE8120 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRE8110 Supplier Promega Supplier No. E8110
Add to Cart
Add to Cart

ONE-Glo™ EX Luciferase Assay System provides both the high sensitivity and long-lived luminescence required to batch-process multiple 96-, 384- or 1,536-well plates. The approximately 2-hour signal half-life provides greater flexibility in assay design.

  • Highly Sensitive Assay with Long-Lived Luminescence
  • Bright, stable signal that can be measured for hours
  • Stable reagent can be stored at room temperature or 4°C
  • Reduced sensitivity to culture media, phenol red and luciferase inhibitors

Specifications

Quantity 10 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.