Learn More
STERIS One Solution st™ Water For Injection
A diluent for surface sterilants, disinfectants, sanitizers, or cleaners used in cleanrooms or other critical environments. Meets USP specifications and is double bagged for easy entry into cleanrooms or aseptic areas.
$604.50 - $623.50
Specifications
Product Type | Water for Injection (WFI) |
---|---|
Sterility | Sterile |
Certifications/Compliance | Meets United States Pharmacopeia (USP) Water For Injection (WFI) Specifications |
Filtered | Filtered |
For Use With (Application) | Diluting Concentrated Cleaners or Disinfectants |
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Price | Quantity & Availability | |||||
19-415-116
|
STERIS
1S0308WR |
Case of 4 for $604.50
|
|
||||||
19-415-117
|
STERIS
1S03E4WR |
2 gal. |
Case of 2 for $623.50
|
|
|||||
Description
- Intended for diluting concentrated cleaners or disinfectants
- Filtered, and filled in a controlled environment
- Each package is double bagged and gamma irradiated for easy entry into controlled environments and aseptic core areas
- Includes an extensive documentation package to meet validation objectives
- Products meet United States Pharmacopeia (USP) Water For Injection (WFI) specifications
- Supported by the STERIS Technical Services team for individualized application and validation assistance
Specifications
Water for Injection (WFI) | |
Meets United States Pharmacopeia (USP) Water For Injection (WFI) Specifications | |
Diluting Concentrated Cleaners or Disinfectants |
Sterile | |
Filtered | |
Double Bagged |