Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ONECUT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 251371AP150UL

Description
Hepatocyte nuclear factor 6 is a CUT homeobox family protein which is a transcription factor and contains a CUT DNA-binding domain and a homeobox DNA-binding domain. It is an upstream regulator of several genes involved in the pathogenesis of maturity-onset diabetes of the young. HNF6 is a ligand of the FOXM1 transcription factor network. HNF6, along with HNF4, controls gene expression in the liver. The expression levels of HNF6 and HNF4 rise steadily during development and that up- or down regulation of HNF6 and HNF4 expression generate premature or delayed expression of the g6pc gene.Specifications
ONECUT1 | |
Polyclonal | |
Unconjugated | |
ONECUT1 | |
Hepatocyte nuclear factor 6, HNF 6, HNF6, HNF6A, One cut domain family member 1, one cut homeobox 1, ONECUT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
15379, 3175 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O08755, Q9UBC0 | |
ONECUT1 | |
ONECUT1 Fusion Protein Ag18250 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction