Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ONECUT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541109
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541109 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541109 Supplier Invitrogen™ Supplier No. PA541109
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NGLHHPGHTQ SHGPVLAPSR ERPPSSSSGS QVATSGQLEE INTKEVAQRI Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

ONECUT2 is a member of the transcription factors of the ONECUT class, whose prototype is hepatocyte nuclear factor (HNF)-6. The distribution of OC-2 mRNA in humans is tissue-restricted, the strongest expression being detected in the liver and skin. ONECUT2 participates in the network of transcription factors required for liver differentiation and metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ONECUT2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ONECUT2
Gene Accession No. O95948
Gene Alias C730009D12; Hepatocyte nuclear factor 6-beta; HNF6B; HNF-6B; HNF-6-beta; OC2; OC-2; One cut domain family member 2; one cut domain, family member 2; one cut homeobox 2; onecut 2; ONECUT2; ONECUT-2 homeodomain transcription factor; RGD1564677; si:ch1073-41k10.1; transcription factor ONECUT2; transcription factor ONECUT-2
Gene Symbols ONECUT2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human ONECUT2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9480
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.