Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ONECUT3 Antibody (10B2F5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237340
Description
ONECUT3 Monoclonal specifically detects ONECUT3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ONECUT3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
OC3, OC-3, one cut domain family member 3, one cut domain, family member 3, one cut homeobox 3, transcription factor ONECUT-3 | |
Mouse | |
50 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
10B2F5 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
ONECUT3 | |
Purified recombinant fragment of human ONECUT3 (AA: 350-455) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
390874 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction