Learn More
Invitrogen™ ONECUT3 Polyclonal Antibody

Description
Positive Control: SW480 cell lysate, Hela cell lysate, PC-3 cell lysate, human colon carcinoma tissue, human stomach tissue, SW620. Subcellular Location: Nucleus.
Specifications
Specifications
| Antigen | ONECUT3 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ONECUT3 |
| Gene Accession No. | O60422, Q8K557 |
| Gene Alias | OC3; OC-3; One cut domain family member 3; one cut domain, family member 3; one cut homeobox 3; Onecut3; Transcription factor ONECUT-3 |
| Gene Symbols | ONECUT3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.