Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ONECUT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566296
Description
Immunogen sequence: SVGHLYGPYGK ELPAMGSPLS PLPN Highest antigen sequence identity to the following orthologs - mouse 92%, rat 52%.
Transcriptional activator. Binds the consensus DNA sequence 5'-DHWATTGAYTWWD-3' on a variety of gene promoters such as those of HNF3B and TTR.
Specifications
ONECUT3 | |
Polyclonal | |
Unconjugated | |
ONECUT3 | |
OC3; OC-3; One cut domain family member 3; one cut domain, family member 3; one cut homeobox 3; Onecut3; Transcription factor ONECUT-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
390874 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60422 | |
ONECUT3 | |
Recombinant Human ONECUT3. Recombinant protein control fragment (Product #RP-106977). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction