Learn More
R&D Systems™ OneStep Blocker Western Blocking Solution & Signal Enhancer
SDP
Catalog No. NB526282 Shop All Novus Products
Change view
Click to view available options
Quantity:
500 mL
Catalog No. Quantity
NB526282 500 mL
1 options
Catalog No. NB526282 Supplier R&D Systems™ Supplier No. NBP333180
Add to Cart
Add to Cart
A Novus product, part of R&D Systems. provides blocking and primary and secondary antibody hybridization in one step and enhances the signal developed with HRP
Specifications
For Use With (Application) Western Blot
Product Line Novus
Quantity 500 mL