Learn More
Specifications
Specifications
| Quantity | 100 Assays |
| Content And Storage | Store at -20°C. |
| Product Line | Novus |
| For Use With (Application) | Label Solution and DAPI (25 X) Should be Stored in The Dark. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
