Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ OneStep TUNEL Apoptosis Kit, Red, 555
SDP

Catalog No. p-200003754
Click to view available options
Quantity:
100 Assays
20 Assays
This item is not returnable. View return policy
This item is not returnable. View return policy

Suitable for in situ apoptosis detection of tissue samples (Paraffin embedding, freezing and ultrathin section) and cells (Cell smears, cell climbing smears).

Specifications

Description OneStep TUNEL
Content And Storage Store at -20C.
Quantity 100 Assays
For Use With (Application) Flow Cytometry, Fluorescence Imaging
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.