Learn More
Dunlop™ Onguard™ Chest Waders with Cleated Outsole
Superior waterproof protection that resists oil, gas, grease, and saltwater in every condition.
Supplier:  Dunlop™ 860670800
 View more versions of this product
Catalog No. 19-024407
Description
Description
These durable chest waders are made of heavy duty PVC and boast cleated outsoles for excellent flexibility and traction clogging prevention, even in wet or muddy terrain.
  • Chest high, with PVC upper and non-absorbent polyester backing
  • Injection molded, seamless boot construction
  • Steel midsole and adjustable polyester and nylon strap
  • Excellent cold weather flexibility
  • Can be folded or rolled without cracking
  • Top drawstring closure for snug fit
  • Non-corrosive, reinforced suspender buttons
  • Built-in belt loops with adjustable belt
  • Replaceable cushion insoles
  • Made in the USA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ASTM F2413-11 M I/75 C/75 SD
Black
PVC, Polyester
8
Polyester/Nylon
Drawstring
Utilities, Municipalities, Aquaculture, and Commercial Fishing
Seamless
Cleated Outsole, Steel Midsole, Replaceable Cushion Insole
Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.