Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dunlop™ Onguard™ Chest Waders with Cleated Outsole

Superior waterproof protection that resists oil, gas, grease, and saltwater in every condition.

$438.60

Specifications

Certifications/Compliance ASTM F2413-11 M I/75 C/75 SD
Closure Type Drawstring
Color Black
For Use With (Application) Utilities, Municipalities, Aquaculture, and Commercial Fishing
Seam Type Seamless
View More Specs

Products 3
Catalog Number Mfr. No. Size Toe Material Price Quantity & Availability  
Catalog Number Mfr. No. Size Toe Material Price Quantity & Availability  
19-073201
Encompass
View Documents
Dunlop™
860671200
12 Steel
Case of 2 for $438.60
Only null left
Add to Cart
 
19-024407
View Documents
Dunlop™
860670800
8 Steel Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area. N/A
19-024406
View Documents
Dunlop™
860671100
11 Steel Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description

Description

These durable chest waders are made of heavy duty PVC and boast cleated outsoles for excellent flexibility and traction clogging prevention, even in wet or muddy terrain.

  • Chest high, with PVC upper and non-absorbent polyester backing
  • Injection molded, seamless boot construction
  • Steel midsole and adjustable polyester and nylon strap
  • Excellent cold weather flexibility
  • Can be folded or rolled without cracking
  • Top drawstring closure for snug fit
  • Non-corrosive, reinforced suspender buttons
  • Built-in belt loops with adjustable belt
  • Replaceable cushion insoles
  • Made in the USA
Specifications

Specifications

ASTM F2413-11 M I/75 C/75 SD
Black
Seamless
Polyester/Nylon
Drawstring
Utilities, Municipalities, Aquaculture, and Commercial Fishing
Cleated Outsole, Steel Midsole, Replaceable Cushion Insole
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.