Learn More
Dunlop™ Onguard™ Men's PVC Overshoes with Four-Way Cleated Outsole
Injection-molded PVC shoes for seamless protection
Supplier: Dunlop™ 88020MD00
This item is not returnable.
View return policy
Description
- Excellent cold weather flexibility
- Four-way, self-cleaning, slip resistant outsole
- Kick off lug for easy on and off
- 10 in. height fits over most work shoes and boots
- Yellow overshoes are made of Flex-O-Thane PVC
Specifications
Yellow | |
10 in. | |
Seamless |
Used in construction, farming, utilities and food processing | |
Flexothane, PVC | |
Medium |