Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dunlop™ Onguard™ Men's PVC Overshoes with Four-Way Cleated Outsole
Injection-molded PVC shoes for seamless protection
$522.24
Specifications
Color | Yellow |
---|---|
For Use With (Application) | Used in construction, farming, utilities and food processing |
Height (English) | 10 in. |
Material | Flexothane, PVC |
Seam Type | Seamless |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-065-520D
![]() |
Dunlop™
88020XL00 |
X-Large |
Case of 6 for $522.24
|
|
|||||
Description
- Excellent cold weather flexibility
- Four-way, self-cleaning, slip resistant outsole
- Kick off lug for easy on and off
- 10 in. height fits over most work shoes and boots
- Yellow overshoes are made of Flex-O-Thane PVC
Specifications
Yellow | |
10 in. | |
Seamless |
Used in construction, farming, utilities and food processing | |
Flexothane, PVC |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title