Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Online Learning Program - 1 Year Class License (1 Teacher/150 Students)
SCI_ED

Catalog No. s98490 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
K to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S98490 K to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S98490 Supplier NewPath Learning™ Supplier No. 510001
Only null left
Add to Cart
Add to Cart

Use interactive, multimedia lessons and innovative teaching tools to engage students through differentiated instruction.

  • Research-based and classroom-tested
  • Innovative and easy-to-use
  • Includes everything needed to provide students with interactive lessons meeting the current state and national standards, including NGSS
  • Engages students and improves their learning through differentiated instruction
  • Allows students to learn anywhere, anytime, on any device (including tablets and Chromebooks)
  • Can also be used on Interactive Whiteboards for classroom instruction
  • Includes the following: click-and-play lessons, narrated multimedia science lessons with interactive exercises, virtual lab investigations, vocabulary, quizzes, and more
  • The program also includes: easy-to-use tools and templates to create and deliver custom lessons to meet individual class and student needs, a unique learning management system to track and analyze student performance, and cloud-based storage to save lessons for future semester or share them with colleagues

Specifications

Grade Level Elementary, Intermediate - Middle School, High School, Higher Ed
Grade K to 14
Category Science, Life Science, Earth Science, Physical Science, Mathematics
Format Download
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.