Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ ONO 2506
GREENER_CHOICESDP

Catalog No. 453050 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-305-0 50 mg
45-301-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-305-0 Supplier R&D Systems™ Supplier No. 4530/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibits S-100β synthesis; neuroprotective

Inhibits S-100β synthesis in activated cultured astrocytes. Prevents delayed infarct expansion 24 hours after permanent middle cerebral artery occlusion (pMCAO) in rats; also exhibits neuroprotective effects in mouse models of Parkinson's disease and Alzheimer's disease.

Chemical Identifiers

CAS 185517-21-9
Molecular Formula C11H22O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 186.29
Purity >98%
Molecular Weight (g/mol) 186.295
Chemical Name or Material ONO 2506
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.