Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ONO AE3 208
GREENER_CHOICESDP

Catalog No. 356550 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-655-0 50 mg
35-651-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-655-0 Supplier Tocris Bioscience™ Supplier No. 3565/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity and selective EP4 antagonist

ONO AE3 208 is a high affinity and selective EP4 receptor antagonist (Ki values are 1.3, 30, 790 and 2400 nM for EP4, EP3, FP and TP receptors respectively). Displays no affinity for EP1, EP2, DP or IP receptors (Ki >10 μM). Inhibits PGE2-induced IL-8 production in colonic epithelial caco-2 cells and attenuates PGE2 inhibition of natural killer T cell activation. Suppresses recovery from experimentally-induced colitis and stimulates CD4+ T cell proliferation in C57BL/6 mice. Also reduces metastasis of mammary tumor cells in a murine model of breast cancer. Orally active.

Specifications

Quantity 50 mg
Product Type ONO AE3 208
Purity >98%
Molecular Weight (g/mol) 404.43
Inhibitors ONO AE3 208

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.