Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OOEP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586033
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586033 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586033 Supplier Invitrogen™ Supplier No. PA586033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OOEP Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen OOEP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene Ooep
Gene Accession No. Q9CWE6
Gene Alias 2410146L05Rik; C6orf156; Factor located in oocytes permitting embryonic development; Floped; HOEP19; KH homology domain containing 2; KH homology domain-containing protein 2; KHDC2; Moep19; OEP19; oocyte and embryo protein 19; oocyte and embryo specific protein; oocyte- and embryo-specific protein 19; oocyte expressed protein; oocyte expressed protein homolog (dog); Oocyte-expressed protein homolog; OOEP; Sddr; STAT3 downstream gene and differentiation regulator
Gene Symbols Ooep
Host Species Rabbit
Immunogen Synthetic peptide within mouse OOEP aa 51-100.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67968
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.