Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OOEP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585954
Description
OOEP Polyclonal Antibody for IHC (P)

Specifications
OOEP | |
Polyclonal | |
Unconjugated | |
Ooep | |
2410146L05Rik; C6orf156; Factor located in oocytes permitting embryonic development; Floped; HOEP19; KH homology domain containing 2; KH homology domain-containing protein 2; KHDC2; Moep19; OEP19; oocyte and embryo protein 19; oocyte and embryo specific protein; oocyte- and embryo-specific protein 19; oocyte expressed protein; oocyte expressed protein homolog (dog); Oocyte-expressed protein homolog; OOEP; Sddr; STAT3 downstream gene and differentiation regulator | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
67968 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
Q9CWE6 | |
Ooep | |
Synthetic peptide within mouse OOEP aa 1-50. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction