Learn More
Eisco™ Oolitic Limestone Sedimentary Rock Specimen
SCI_ED
Catalog No. S27610 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1/Ea.
12/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S27610 12/Pk.
S27588 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S27610 Supplier Eisco™ Supplier No. ESNG0081PK12
Add to Cart
Add to Cart
Oolitic limestone is a carbonate rock formed from the cementation of thousands of ooids, tiny nodules formed from the accumulation of carbonate material around a nucleus. Oolitic limestone forms in reef or lagoon environments.
  • Correlates well to Next Generation Science Standards Evidence
  • Useful in the classroom for lessons on depositional facies, sedimentary rocks, and stratigraphy
Specifications
Product Type Sedimentary Rock
Grade K to 16
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.1 kg