Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OOSP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549127
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549127 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549127 Supplier Invitrogen™ Supplier No. PA549127
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OOSP1 Polyclonal Antibody for Western Blot

May be involved in cell differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OOSP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OOSP1
Gene Accession No. A8MZH6
Gene Alias oocyte secreted protein 1, pseudogene; OOSP1
Gene Symbols OOSP1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 82-108 amino acids from the C-terminal region of human OOSP1.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255649
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.