Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OP Pesticide Calibration Mix A, U.S. EPA Method 8140/8141, Restek
SDP

Catalog No. 06719893 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-893 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-893 Supplier Restek Corporation Supplier No. 32277
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Organophosphorous Pesticides

Specifications

Analyte or Component Names 20 components:
azinphos methyl
bolstar (sulprofos)
chlorpyrifos
coumaphos
demeton O and S
diazinon
dichlorvos
disulfoton
ethoprophos
fensulfothion
fenthion
merphos
methyl parathion
mevinphos
naled
phorate
ronnel
stirofos
tokuthion
(prothiofos)
trichloronate
Purity Grade Notes Determined by a combination of methods: GC/FID, GC/FPD, GC/NPD, DSC, and HPLC/UV
Physical Form Volatile
Element Azinphos Methyl, Bolstar (sulprofos), Chlorpyrifos, Coumaphos, Demeton O+S, Diazinon, Dichlorvos, Disulfoton, Ethoprophos, Fensulfothion, Fenthion, Merphos, Methyl Parathion, Mevinphos, Naled, Phorate, Ronnel, Stirofos, Tokuthion, Prothiofos, Trichloronate
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 200μg/mL each in hexane:acetone (95:5)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.