Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OP Pesticide Calibration Mix B, U.S. EPA Method 8141A, Restek
SDP

Catalog No. 06719894 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-894 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-894 Supplier Restek Corporation Supplier No. 32278
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material OP Pesticide Calibration Mix B
Name Note U.S. EPA Method 8141A
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 200μg/mL each in hexane:acetone (95:5):
Dimethoate
EPN
Malathion
Monocrotophos
Parathion
Sulfotepp
TEPP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.