Learn More
OPA1 Antibody (1284B), HRP, Novus Biologicals™
SDP
Rabbit Monoclonal Antibody
Supplier:  Novus Biologicals NBP259770H
Catalog No. NBP259770H
Only null left
Add to Cart
Description
Description
OPA1 Monoclonal specifically detects OPA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
OPA1
Monoclonal
HRP
PBS with No Preservative
OPA1
A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot# O60313]
0.1 mL
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
1284B
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1
Rabbit
Protein A or G purified
Apoptosis, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Sensory Systems, Vision
4976
Store at 4C in the dark.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.